Bulb pipettes, Qualicolor, colour coded (one mark), Class B
Supplier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N632433131014 N632433131016 N632433131018 N632433131019 N632433131022 N632433131023 632433131025 632433131030 632433131037
KAVAN632433131014EA 10.4 GBP
KAVAN632433131014 KAVAN632433131016 KAVAN632433131018 KAVAN632433131019 KAVAN632433131022 KAVAN632433131023 KAVA632433131025 KAVA632433131030 KAVA632433131037
Bulb pipettes, Qualicolor, colour coded (one mark), Class B
Pipettes Bulb Pipettes
Soda-lime glass.
  • Enlarged form
  • Not autoclavable at 121 °C
Certifications: DIN 12690, ISO 648