Volumetric flasks for sugar analysis
Supplier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
632431510030 632431510037
KAVA632431510030EA 89.6 GBP
KAVA632431510030 KAVA632431510037
Volumetric flasks for sugar analysis
Flasks Volumetric Flasks
Borosilicate glass, clear.
  • With two graduation marks
  • White markings
  • Autoclavable at 121 °C