Flasks, pear- shaped, with standard ground joint glass stopper
Supplier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
632426372250 632426372025
KAVA632426372250EA 710 GBP
KAVA632426372250 KAVA632426372025
Flasks, pear- shaped, with standard ground joint glass stopper
Flasks Pear Shaped Flasks
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.