Weighing Bottles, Glass
Supplier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
632421202505 632421202343 632421202604 632421202605 632421202804 632421202503
KAVA632421202505EA 1560 GBP
KAVA632421202505 KAVA632421202343 KAVA632421202604 KAVA632421202605 KAVA632421202804 KAVA632421202503
Weighing Bottles, Glass
Bottles Weighing Bottles
Made of borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance