Desiccators with 24/29 and stopcock, vacuum rated
Supplier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
632415262201 632415272150 632415272300 632415272250 632415262151 632415262101 632415262311
KAVA632415262201EA 170 GBP
KAVA632415262201 KAVA632415272150 KAVA632415272300 KAVA632415272250 KAVA632415262151 KAVA632415262101 KAVA632415262311
Desiccators with 24/29 and stopcock, vacuum rated
Desiccators Desiccators, Glass
SIMAX borosilicate glass.
  • Ideal for storing samples which are sensitive to humidity
  • Suitable for a range of tasks under vacuum
  • All parts are interchangeable and replacement parts are available in the event of breakage
  • Available with or without plate