Desiccators, with porcelain plate
Supplier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
632415270150 632415270300 632415270250 632415271200 632415270100 632415271300 632415271150
KAVA632415270150EA 441 GBP
KAVA632415270150 KAVA632415270300 KAVA632415270250 KAVA632415271200 KAVA632415270100 KAVA632415271300 KAVA632415271150
Desiccators, with porcelain plate
Desiccators Desiccators, Glass
Simax borosilicate glass, novus pattern.
  • Vacuum or knob lid type with tubular in centre of lid
  • Cover ground to fit
  • Spare lids are available for these desiccators
Delivery information: Supplied with lid, removable stopcock (vacuum type only) and perforated ceramic disc.