Automatic burettes, Schilling pattern, Class B
Supplier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1980 1981
KART1980EA 252 GBP
KART1980 KART1981
Automatic burettes, Schilling pattern, Class B
Burettes
PMP/PE, class B.
  • Push button for drop-by- drop delivery
  • PE reservoir with stable LDPE base
According to DIN 12.700 standards, class B
Certifications: According to DIN 12.700 standards, class B