Industrial scoops, Plastilab®
Supplier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1066 1111 1110 1112
KART1066EA 220 GBP
KART1066 KART1111 KART1110 KART1112
Industrial scoops, Plastilab®
Scoops
These scoops made from HDPE are designed for heavy industrial use.
  • Round shaped design facilitates complete transfer of sampling material
  • Suitable for foodstuff