Tyvek GMP Equipment Cover
Supplier: KEYSTONE CLEANROOM PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
COVTVK602668WS
KACCCOVTVK602668WSEA 4860 GBP
KACCCOVTVK602668WS
Tyvek GMP Equipment Cover
Bags Sterilisation Pouches
Save time and standardise the activities in equipment preparation for steam sterilisation; or use in any pharmaceutical manufacturing setting to keep cleaned equipment protected from contamination.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Individually poly bagged and labelled
  • Steam sterilisation indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
These covers provide a convenient, time-saving method of wrappingMore Product Information