Plant RNA purification Mini kit, GeneJET™
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
K0801 K0802
K0801EA 374 GBP
K0801 K0802
Plant RNA purification Mini kit, GeneJET™
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
GeneJET™ Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from a wide variety of plant species and tissue types.
  • Fast - procedure completed in 20 minutes following cell lysis
  • Convenient - protocol does not include clogging-prone lysate filtration step
  • High yield - up to 65 µg of high purity and high integrity RNA from 50 mg plant sample
  • Compatible with wide variety of samples (leaves, roots, sprouts and other parts of various plants)
The kit utilises silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample.
Delivery information: Each kit includes Plant RNA Lysis Solution, Wash Buffer WB 1 (concentrated), Wash Buffer 2 (concentrated), water (nuclease-free), GeneJET™ RNA purification columns (pre-assembled with collection tubes), collection tubes (1,5 and 2 ml).