Whole blood genomic DNA purification mini kit, GeneJET™
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
K0781 K0782
K0781EA 189 GBP
K0781 K0782
Whole blood genomic DNA purification mini kit, GeneJET™
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
GeneJET™ Whole Blood Genomic DNA Purification Mini Kit utilises silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting, and enzymatic reactions.
  • Efficient: High yields of high molecular weight genomic DNA
  • Fast: 20 minute procedure following cell lysis
  • Convenient: Spin columns are capped and assembled with collection tubes
  • Pure: DNA pure from enzyme inhibitors, RNA and proteins ready to use for all demanding applications; validated in real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests
  • Compatible: Suitable for wide variety of samples like fresh or frozen anticoagulated mammalian blood (treated with EDTA, citrate or heparin), aged blood, buffy coat, bone marrow, and body fluids
Delivery information: Each kit includes Proteinase K solution, lysis solution, Wash Buffer I (concentrated), Wash Buffer II (concentrated), elution buffer, GeneJET™ purification columns (pre-assembled with collection tubes), and collection tubes.