Safety dispensing cans, HDPE
Supplier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
14002 14004 14010
JUSU14002EA 58.2 GBP
JUSU14002 JUSU14004 119-0275
Safety dispensing cans, HDPE
Safety Cans
These FM approved brass valves are ideal for use with flammables.
HDPE, textured.
  • Permit easy, one-hand dispensing of liquids directly onto work surface
  • Leakproof, self-closing, brass dispenser valve minimises spills and controls excess
Order Now
SPECIFICATIONS
Environmentally Preferable