Temperature control systems, PRESTO®
Supplier: JULABO GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9420300 9420401 9421401 9420801 9421801 9420801.T 9421801.T 9421912 9421923 9421913 9421922 9421912.TT 9421913.TT 9421922.TT 9421923.TT 9420452 9420452.T 9420852 9420852.T 9421852 9421852.T 9421502.T 9421502 9421552 9421562 9421563.S1 9420381.N1 9420411.N1 9420701.N1 9421411.N1 9421932.N1 9421933.N1 9421583.S1
461-0635EA 12344 GBP
461-0635 461-0636 461-0637 461-0735 461-0736 461-0879 461-0880 461-0804 461-0813 461-0807 461-0810 461-0806 461-0809 461-0812 461-0815 461-0881 461-0882 461-1107 461-1108 461-1109 461-1110 461-1006 461-1005 JULA9421552 JULA9421562 JULA9421563.S1 JULA9420381.N1 JULA9420411.N1 JULA9420701.N1 JULA9421411.N1 JULA9421932.N1 JULA9421933.N1 JULA9421583.S1
Temperature control systems, PRESTO®
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
PRESTO® systems cover a working temperature range of −93 to +250 °C with high cooling and heating capacity. These instruments can compensate for exothermic and endothermic reactions with extraordinary speed, they feature high flow rates, constant pressure and a controlled build-up of pump pressure. Changes in the temperature control of liquid viscosity are balanced dynamically. Permanent internal monitoring and self-lubricating pumps contribute to long service life. The integrated touch panel gives the user a clear view of important information. Fingertip control via the touch screen.More Product Information