MAGIO™ MX Refrigerated/Heating Circulators with Natural Refrigerant
Supplier: JULABO GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9033751.N1 9033752.N1
JULA9033751.N1EA 13100 GBP
JULA9033751.N1 JULA9033752.N1
MAGIO™ MX Refrigerated/Heating Circulators with Natural Refrigerant
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
The MAGIO™ MX refrigerated/heating circulators operate with a working temperature range of −50 to +200 °C and heating capacity of 3 kW. With their powerful pressure/suction pump the MAGIO™ units are ideal for external temperature applications including reactors to 30 L. An extensive range of accessories is available.
  • Integrated programmer allows temperature time profiles to be run
  • Stainless steel wetted parts
  • R1270 natural refrigerant
  • Bright, easy to read touchscreen display
  • Variable speed compressors reduce energy consumption and lower noise
  • Powerful,
More Product Information