Chromatography tubing, FEP
Supplier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-T-6320-M3 JR-T-6105-M10 JR-T-6105-M25 JR-T-6105-M3 JR-T-6210-M10 JR-T-6210-M25 JR-T-6210-M3 JR-T-6310-M10 JR-T-6310-M25 JR-T-6310-M3 JR-T-6320-M10 JR-T-6605C-M10 JR-T-6620C-M10 JR-T-6620C-M25 JR-T-6620C-M3 JR-T-6620-M1 JR-T-6620-M10 JR-T-6620-M3 JR-T-6615-M10 JR-T-6615-M1 JR-T-6606-M3 JR-T-6605C-M3 JR-T-6605-M1 JR-T-6605-M10 JR-T-6605-M3 JR-T-6606C-M10 JR-T-6606C-M25 JR-T-6606C-M3 JR-T-6615-M3 JR-T-6606-M10 JR-T-6607C-M10 JR-T-6607C-M25 JR-T-6607C-M3 JR-T-6607-M1 JR-T-6607-M10 JR-T-6607-M3 JR-T-6610C-M10 JR-T-6610-M1 JR-T-6610C-M25 JR-T-6610C-M3 JR-T-6610-M10 JR-T-6610-M3 JR-T-6606-M1
JOURJR-T-6320-M3EA 128 GBP
JOURJR-T-6320-M3 JOURJR-T-6105-M10 JOURJR-T-6105-M25 JOURJR-T-6105-M3 JOURJR-T-6210-M10 JOURJR-T-6210-M25 JOURJR-T-6210-M3 JOURJR-T-6310-M10 JOURJR-T-6310-M25 JOURJR-T-6310-M3 JOURJR-T-6320-M10 JOURJR-T-6605C-M10 JOURJR-T-6620C-M10 JOURJR-T-6620C-M25 JOURJR-T-6620C-M3 JOURJR-T-6620-M1 JOURJR-T-6620-M10 JOURJR-T-6620-M3 JOURJR-T-6615-M10 JOURJR-T-6615-M1 JOURJR-T-6606-M3 JOURJR-T-6605C-M3 JOURJR-T-6605-M1 JOURJR-T-6605-M10 JOURJR-T-6605-M3 JOURJR-T-6606C-M10 JOURJR-T-6606C-M25 JOURJR-T-6606C-M3 JOURJR-T-6615-M3 JOURJR-T-6606-M10 JOURJR-T-6607C-M10 JOURJR-T-6607C-M25 JOURJR-T-6607C-M3 JOURJR-T-6607-M1 JOURJR-T-6607-M10 JOURJR-T-6607-M3 JOURJR-T-6610C-M10 JOURJR-T-6610-M1 JOURJR-T-6610C-M25 JOURJR-T-6610C-M3 JOURJR-T-6610-M10 JOURJR-T-6610-M3 JOURJR-T-6606-M1
Chromatography tubing, FEP
Tubing Chromatography Tubing
FEP, natural or coloured.
  • Solid colours identify the tubing Int.Ø.
  • Recommend for ion chromatography applications
FEP (FluorinatedEthylenePropylene) tubing is chemically inert to most HPLC solvents, and is recommended for low pressure applications.