Chromatography tubing, PFA
Supplier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-T-4001-M3 JR-T-4002-M3 JR-T-4003-M3 JR-T-4007-M3 JR-T-4007-M25 JR-T-4007-M10 JR-T-4003-M25 JR-T-4003-M10 JR-T-4002-M10 JR-T-4001-M25 JR-T-4001-M10 JR-T-4002-M25 JR-T-4004
JOURJR-T-4001-M3EA 108 GBP
JOURJR-T-4001-M3 JOURJR-T-4002-M3 JOURJR-T-4003-M3 JOURJR-T-4007-M3 JOURJR-T-4007-M25 JOURJR-T-4007-M10 JOURJR-T-4003-M25 JOURJR-T-4003-M10 JOURJR-T-4002-M10 JOURJR-T-4001-M25 JOURJR-T-4001-M10 JOURJR-T-4002-M25 JOURJR-T-4004
Chromatography tubing, PFA
Tubing Chromatography Tubing
These PFA (perfluoro alkoxy) tubing has excellent chemical stability and mechanical strength.
  • Maximum pressure rating of up to 100 bar