Safety exhaust filters
Supplier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-S-20002 JR-S-20005
JOURJR-S-20002EA 173 GBP
JOURJR-S-20002 JOURJR-S-20005
Safety exhaust filters
Filters Venting Filters
These safety exhaust filters can be placed on the waste bottle of an HPLC system, to protect workers from solvent vapors in the ambient air.
  • Prevents pollution of ambient air with volatile compounds
  • Break-through detector alerts the users about the saturation of the filter absorbent
  • Universal 1/4-28 connection
The break-through detector recognises trace amounts of acetone, acetonitrile, ethanol, ethyl acetate, methanol, methylene chloride and THF. When the absorbent is saturated, break-through is indicated by a colour change in the detector fromMore Product Information