Mobile phase filters
Supplier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-M3678-25
JOURJR-M3678-25EA 0 GBP
JOURJR-M3678-25
Mobile phase filters
Filters Liquid Process Filters
These filters made from stainless steel 316L protect HPLC systems from small particles in the mobile phase.
  • Suitable for most solvents
  • Version for both analytical and preparative applications