Precolumn filters, high pressure
Supplier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-68262 JR-68258 JR-68264 JR-68264PE JR-1151-10P-5
JOURJR-68262EA 213 GBP
JOURJR-68262 JOURJR-68258 JOURJR-68264 JOURJR-68264PE JOURJR-1151-10P-5
Precolumn filters, high pressure
Filters Precolumn Filters and Frits
Precolumn filter kit has a standard 10 to 32 fitting and fits direct to most column types without introducing an additional dead volume.
  • Minimum hold-up volume
  • Metal free PE frits and biocompatible titanium frits
It protects columns against fine particles, which may otherwise accumulate on the column frit, leading to split peaks and high backpressure.