Low pressure tees and crosses, PEEK
Supplier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-6030 JR-6031 JR-6032 JR-6033 JR-6040 JR-6041 JR-6042 JR-6043 JR-6044 JR-6039 JR-6034 JR-6029 JR-B-C5M1PK JR-B-C9M2PK JR-B-C5M2PK JR-B-C9M1PK
JOURJR-6030EA 189 GBP
JOURJR-6030 546-1343 JOURJR-6032 JOURJR-6033 JOURJR-6040 JOURJR-6041 JOURJR-6042 JOURJR-6043 JOURJR-6044 JOURJR-6039 JOURJR-6034 JOURJR-6029 JOURJR-B-C5M1PK JOURJR-B-C9M2PK JOURJR-B-C5M2PK JOURJR-B-C9M1PK
Low pressure tees and crosses, PEEK
Fittings and Connectors
Connectors for low pressure applications are machined from PEEK.
  • Flat-bottomed 1/4-28 fitting details
  • Completely inert and biocompatible
  • Available as the body only, or complete with fittings