Fingertight adapters, Color-It
Supplier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-55015-5 JR-55016-12 JR-55014-5 JR-55013-5 JR-55012-5 JR-55011-5 JR-55010-5
JOURJR-55015-5EA 15.3 GBP
JOURJR-55015-5 JOURJR-55016-12 JOURJR-55014-5 JOURJR-55013-5 JOURJR-55012-5 JOURJR-55011-5 JOURJR-55010-5
Fingertight adapters, Color-It
Fittings and Connectors
These fingertight PP adapters are used with ¹/₄" PEEK and stainless hex-head nuts, converting them into colour coded fingertight fittings.
  • Fit all hex-head PEEK and SS fittings for ¹/₁₆" tubing
  • Used in low and high pressure
  • Available in six colours for easy solvent line identification