Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J67802.AP J67802.K2
J67802.APEA 29.9 GBP
J67802.AP J67802.K2
Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium
Buffers Cell Culture Buffers

Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00131855

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR