Krebs-Ringer solution, HEPES-buffered

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J67795.AP
J67795.APEA 103 GBP
J67795.AP
Krebs-Ringer solution, HEPES-buffered
Buffers Cell Culture Buffers

Contains 120 mM sodium chloride, 5 mM potassium chloride, 2 mM calcium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, 5,5 mM HEPES, and 1 mM D-glucose


  • pH 7,2
  • Prepared in 18,2 megohms water
  • Filtered through 0,22 µm filter


Physical Form: Liquid
Appearance: Colourless


Standard physiological buffer that is suitable for various calcium imaging experiments. It is also used as a preserving agent.

Order Now

SPECIFICATIONS

Environmentally Preferable
Appearance Colourless
Physical form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR