Gold 95% supplied in 0.1 mM PBS, nanoparticles 150 nm

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J67276.ACEA 92.7 GBP
J67276.AC J67276.AE
Gold 95% supplied in 0.1 mM PBS, nanoparticles 150 nm
Gold

Purity: 95% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Dark brown to pink MDL No.: MFCD00003436


Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

Formula: Au
Storage Temperature: Refrigerator
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR