Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 20 nm (Abs. 405 nm)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J67067.ACEA 68.5 GBP
J67067.AC J67067.AE
Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 20 nm (Abs. 405 nm)
Silver

CAS No.: 7440-22-4 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Yellow Odour: Odourless MDL No.: MFCD00003397

Formula: Ag
Storage Temperature: Refrigerator
MDL Number: MFCD00003397
CAS Number: 7440-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR