Calcium-D-saccharate tetrahydrate 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Calcium D-glucarate tetrahydrate

J66947.14 J66947.22
J66947.14EA 30 GBP
J66947.14 J66947.22
Calcium-D-saccharate tetrahydrate 98%
Calcium-D-saccharate tetrahydrate

D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.


  • Soluble in water( 0,43 g/l)


Caution: Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Formula: C₆H₈CaO₈.4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150704
CAS Number: 5793-89-5
EINECS: 227-334-1
Merck Index: 14,1700

Order Now

Specification Test Results

Colour White
Form Powder

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR