n-Dodecyl-β-D-maltoside

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

J66869.ME J66869.03 J66869.06
J66869.MEEA 84.3 GBP
J66869.ME J66869.03 J66869.06
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
Formula: C₂₄H₄₆O₁₁
Melting Pt: 224…226 °C
Storage Temperature: Freezer
MDL Number: MFCD00043012
CAS Number: 69227-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR