4-Methylumbelliferyl-β-D-glucopyranoside 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-Methylumbelliferyl beta-D-Glucopyranoside, 4-Methylumbelliferyl β-D-Glucopyranoside, 4-MU-β-D-Glc, 4-Methyl-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, MUGlucosid

J66630.MD J66630.03 J66630.06
J66630.MDEA 32.6 GBP
J66630.MD J66630.03 J66630.06
4-Methylumbelliferyl-β-D-glucopyranoside 99%
4-Methylumbelliferyl-beta-D-glucopyranoside
Formula: C₁₆H₁₈O₈
Melting Pt: 210…212 °C
Storage Temperature: Freezer
MDL Number: MFCD00063694
CAS Number: 18997-57-4
EINECS: 242-736-7

Order Now

Specification Test Results

Solubility Soluble in DMF (Dimethylformamide) or water
Purity 99%
Physical form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR