Itraconazole 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Oriconazole

J66390.06 J66390.14
J66390.06EA 108 GBP
J66390.06 J66390.14
Itraconazole 98%
Itraconazole

Purity: 98% CAS No.: 84625-61-6 Molecular Formula: C35H38Cl2N8O4 Formula Weight: 705.63 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White to off-white Melting Point: 166° Solubility: Insoluble in water. Solube in chloroform at 50 mg/ml. Slightly soluble in ethanol or methanol MDL No.: MFCD00870168

Formula: C₃₅H₃₈Cl₂N₈O₄
Melting Pt: 166 °C
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00870168
CAS Number: 84625-61-6
Merck Index: 14,5245

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR