Cefmetazole sodium 921 µg/mg

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

J66348.03
J66348.03EA 77.7 GBP
J66348.03
Cefmetazole sodium 921 µg/mg
Cefmetazole sodium

Cefmetazole is used to study the effect of expression, binding, and inhibition of penicillin-binding proteins (with the exception of PBP2) on bacterial cell wall mucopeptide synthesis. It is also used to study protein mediated antibiotic transport.


CAS No.: 56796-39-5 Molecular Formula: C15H16N7O5S3Na Formula Weight: 493.52 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white Solubility: Soluble in water MDL No.: MFCD00214260


Applications: Cefmetazole is used to study the effect of expression, binding, and inhibition of penicillin-binding proteins (with the exception of PBP2) on bacterial cell wall mucopeptide synthesis. It is also used to study protein mediated antibiotic transport.

Formula: C₁₅H₁₆N₇NaO₅S₃
MDL Number: MFCD00214260
CAS Number: 56796-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR