Cefprozil

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J66326.MCEA 106 GBP
J66326.MC
Cefprozil
Cefprozil

CAS No.: 92665-29-7 Molecular Formula: C18H19N3O5S Formula Weight: 389.42 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White Melting Point: 218-225° Boiling Point: 803° Solubility: Soluble in DMSO at 10mg/ml. Insoluble in water. MDL No.: MFCD00865082

Formula: C₁₈H₁₉N₃O₅S
MW: 389.426 g/mol
Boiling Pt: 803 °C
Melting Pt: 218…225 °C
Flash Pt: >110 °C (230 °F)
MDL Number: MFCD00865082
CAS Number: 92665-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR