Nonyl-β-D-glucopyranoside ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J66009.03 J66009.06
J66009.03EA 159 GBP
J66009.03 J66009.06
Nonyl-β-D-glucopyranoside ≥98%
Nonyl β-D-glucopyranoside
Formula: C₁₅H₃₀O₆
Storage Temperature: Freezer
MDL Number: MFCD00063300
CAS Number: 69984-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR