Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J63677.K2 J63677.K3 J63677.K7
J63677.K2EA 47.3 GBP
J63677.K2 J63677.K3 J63677.K7
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Buffers Electrophoresis Buffers Running Buffers

Physical Form: Liquid
MDL No.: MFCD00236357

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR