Buffers, PIPES, 0,5 M solutions

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J61786.AE J61786.AK J61224.AE J61224.AK J61406.AE J61406.AK J63234.AE J63234.AK J63617.AE J63617.AK
J61786.AEEA 70.3 GBP
J61786.AE J61786.AK J61224.AE J61224.AK J61406.AE J61406.AK J63234.AE J63234.AK J63617.AE J63617.AK
Buffers, PIPES, 0,5 M solutions
Buffers Biological Buffers

Liquid


CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159


Caution: Storage Temperature: 2 to 8 °C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR