Captopril

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (S)-(-)-1-(3-Mercapto-2-methyl-1-oxopropyl)-L-proline, (S)-(-)-1-(3-Mercapto-2-methylpropionyl)-L-proline, (S)-1-((S)-3-mercapto-2-methylpropanoyl)pyrrolidine-2-carboxylic acid

J63593.03 J63593.06 J63593.14
J63593.03EA 41.6 GBP
J63593.03 J63593.06 J63593.14
Captopril
Captopril

CAS No.: 62571-86-2 Molecular Formula: C9H15NO3S Formula Weight: 217.29 Physical Form: Powder Appearance: White to off-white Melting Point: 104-108° Beilstein Reference: 477887 MDL No.: MFCD00168073


Applications: Orally active inhibitor of angiotensin-converting enzyme

Formula: C₉H₁₅NO₃S
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00168073
CAS Number: 62571-86-2
EINECS: 263-607-1
Merck Index: 14,1774

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR