Thiamphenicol

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-[4-(methylsulphonyl)phenyl]-2-propyl]acetamide, (Methylsulphonyl)chloramphenicol

J63575.03 J63575.06 J63575.14
J63575.03EA 22.8 GBP
J63575.03 J63575.06 J63575.14
Thiamphenicol
Thiamphenicol

CAS No.: 15318-45-3 Molecular Formula: C12H15Cl2NO5S Formula Weight: 356.22 Physical Form: Powder Appearance: White to yellow Solubility: Soluble in acetonitrile or DMF. Slightly soluble in water Beilstein Reference: 2819542 MDL No.: MFCD00467983


Applications: The methyl-sulfonyl analogue of chloramphenicol

Formula: C₁₂H₁₅Cl₂NO₅S
MW: 356.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00467983
CAS Number: 15318-45-3
EINECS: 239-355-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR