Genistein ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone

J63241.MC J63241.03 J63241.09
J63241.MCEA 23.5 GBP
J63241.MC J63241.03 J63241.09
Genistein ≥99%
Genistein

4',5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.


Purity: 99+% CAS No.: 446-72-0 Molecular Formula: C15H10O5 Formula Weight: 270.24 Storage Temperature: -30°C to -10°C Physical Form: Powder Melting Point: ca 300° Beilstein Reference: 263823 Merck Reference: 14,4391 MDL No.: MFCD00016952


Inhibitor of tyrosine protein kinase

Formula: C₁₅H₁₀O₅
Melting Pt: ∼ 300 °C
MDL Number: MFCD00016952
CAS Number: 446-72-0
EINECS: 207-174-9
Merck Index: 14,4391

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR