Manganese(II) chloride 1 M in aqueous solution

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Manganese dichloride

J63150.ADEA 45.7 GBP
J63150.AD J63150.AE
Manganese(II) chloride 1 M in aqueous solution
Manganese (II) chloride

CAS No.: 7773-01-5 Molecular Formula: Cl2Mn Formula Weight: 125.84 Physical Form: Liquid Appearance: Colourless

Formula: MnCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR