Orlistat 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N-Formyl-L-leucine (1S)-1-[[(2S,3S)-3-Hexyl-4-oxo-2-oxetanyl]methyl]dodecyl Ester, (S)-(S)-1-((2S,3S)-3-Hexyl-4-oxooxetan-2-yl)tridecan-2-yl 2-formamido-4-methylpentanoate

J62999.MF J62999.MC J62999.MD
J62999.MFEA 61.1 GBP
J62999.MF J62999.MC J62999.MD
Orlistat 98%
Orlistat

Purity: 98% CAS No.: 96829-58-2 Molecular Formula: C29H53NO5 Formula Weight: 495.73 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: White Melting Point: 50° MDL No.: MFCD05662360


Applications: Novel inhibitor of fatty acid sythase. Halts tumor cell proliferation and induces tumor cell apoptosis

Formula: C₂₉H₅₃NO₅
MW: 495.75 g/mol
MDL Number: MFCD05662360
CAS Number: 96829-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR