Phosphate-buffered saline (PBS, 10X), RNase free

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J62851.AK J62851.AP
J62851.AKEA 82.4 GBP
J62851.AK J62851.AP
Phosphate-buffered saline (PBS, 10X), RNase free
Buffers Cell Culture Buffers

Physical Form: Liquid
MDL No.: MFCD00131855

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR