GTE buffer

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J62597.AE J62597.AK
J62597.AEEA 29.7 GBP
J62597.AE J62597.AK
GTE buffer
Buffers Extraction Buffers

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Applications: For miniprep and DNA purification

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR