Trichloroacetic acid (TCA) 5% (w/v) in aqueous solution

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

J62366.K2 J62366.AP
J62366.K2EA 79.6 GBP
J62366.K2 J62366.AP
Trichloroacetic acid (TCA) 5% (w/v) in aqueous solution
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 2564
ADR: 8,III
Merck Index: 14,9627
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR