TRIS-glycine native sample buffer (4X), pH 8,6

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J61864.ACEA 52 GBP
J61864.AC J61864.AD
TRIS-glycine native sample buffer (4X), pH 8,6
Buffers Electrophoresis Buffers Loading Buffers

Physical Form: Liquid
Appearance: Colourless to yellow
MDL No.: MFCD00236360
Applications: For preparation of protein samples for native gel electrophoresis

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR