Sodium pyruvate Cell Culture

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

J61840.18 J61840.22
J61840.18EA 102 GBP
J61840.18 J61840.22
Sodium pyruvate Cell Culture
Sodium pyruvate

CAS No.: 113-24-6 Molecular Formula: C3H3NaO3 Formula Weight: 110.04 Physical Form: Powder Appearance: White to pale yellow Solubility: Freely soluble in water Beilstein Reference: 3568341 MDL No.: MFCD00002586

Formula: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR