β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form), NADH disodium salt, ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt

J61638.03 J61638.06
J61638.03EA 54.1 GBP
J61638.03 J61638.06
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) 97%
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)

Purity: 98% CAS No.: 606-68-8 Molecular Formula: C21H27N7Na2O14P2 Formula Weight: 709.41 Storage Temperature: 2°C to 8°C Physical Form: Powder Beilstein Reference: 5230241 MDL No.: MFCD00036200

Formula: C₂₁H₂₇N₇Na₂O₁₄P₂
Storage Temperature: Refrigerator
MDL Number: MFCD00036200
CAS Number: 606-68-8
EINECS: 210-123-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR