Aluminium chloride 0.5 mol/l in aqueous solution

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Aluminum chloride

J61582.AEEA 49.2 GBP
J61582.AE
Aluminium chloride 0.5 mol/l in aqueous solution
Aluminium chloride
Formula: AlCl₃
MW: 241.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 2581
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR