Mevastatin 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J61357.X0 J61357.MA J61357.MB
J61357.X0EA 57.6 GBP
J61357.X0 J61357.MA J61357.MB
Mevastatin 98%
Mevastatin

Inhibitor of HMG-CoA reductase. Induces apoptosis.


Applications: Inhibitor of HMG-CoA reductase. Induces apoptosis

Formula: C₂₃H₃₄O₅
Storage Temperature: Refrigerator
MDL Number: MFCD05662341
CAS Number: 73573-88-3
Merck Index: 14,6164

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR