Dextran, M.W. ~ 20,000

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J61216.09 J61216.18 J61216.22 J61216.36
J61216.09EA 21.9 GBP
J61216.09 J61216.18 J61216.22 J61216.36
Dextran, M.W. ~ 20,000
Dextran

CAS No.: 9004-54-0 Molecular Formula: (C6H10O5)n Physical Form: Powder Appearance: White to off-white Merck Reference: 14,2948 MDL No.: MFCD00130935

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5
Merck Index: 14,2948

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR