Dacarbazine

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 5- (3,3-Dimethyl-1-triazenyl) imidazole-4-carboxamide, DTIC, 5-(3,3-Dimethyltriazeno)imidazole-4-carboxamide

J61023.ME J61023.03
J61023.MEEA 101 GBP
J61023.ME J61023.03
Dacarbazine
Dacarbazine

CAS No.: 4342-03-4 Molecular Formula: C6H10N6O Formula Weight: 182.18 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale yellow Melting Point: 210-212° Solubility: Slightly soluble in water, DMSO and ethanol MDL No.: MFCD00057167


Applications: Used in treatment of malignant melanoma and sarcomas. Generates methyl adducts to purine bases in DNA

Formula: C₆H₁₀N₆O
MW: 182.19 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD00057167
CAS Number: 4342-03-4
EINECS: 224-396-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR