TRIS-buffered saline (TBS, 10X) pH 7,4

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

J60764.K2 J60764.K3 J60764.K7
J60764.K2EA 50.8 GBP
J60764.K2 J60764.K3 J60764.K7
TRIS-buffered saline (TBS, 10X) pH 7,4
Buffers Electrophoresis Buffers Washing Buffers

Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00132476

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR